Anti-CENPE Mouse Monoclonal Antibody [clone: 1H12]

Lieferant: Abnova
ABNOMAB1924EA 904 EUR
ABNOMAB1924
Anti-CENPE Mouse Monoclonal Antibody [clone: 1H12]
Antikörper
Mouse monoclonal antibody raised against full length recombinant.

Recommended Dilutions: Western Blot: 1:500-1:2000; Immunoprecipitation: 1:200; Immunofluorescence: 1:500-1:2000

Type: Primary
Antigen: CENPE
Clonality: Monoclonal
Clone: 1H12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR