Anti-FITC Monoclonal Antibody (Biotin) [clone: NI 239]

Lieferant: Abnova
ABNOMAB10078EA 643 EUR
ABNOMAB10078
Anti-FITC Monoclonal Antibody (Biotin) [clone: NI 239]
Antikörper
Mouse monoclonal antibody raised against FITC.

Recommended Dilutions: ELISA: 1:500; Western Blot: 1:1000; Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: FITC
Clonality: Monoclonal
Clone: NI 239
Conjugation: Biotin
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR