Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4G7]

Lieferant: Abnova
ABNOMAB10234EA 399 EUR
ABNOMAB10234
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4G7]
Antikörper
Mouse monoclonal antibody raised against of CD19.

Recommended Dilutions: Flow Cytometry: 20 ?l reagent/100 ?l of whole blood or 10^6 cells

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 4G7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR