Anti-EPCAM Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: ESA 214]

Lieferant: Abnova
ABNOMAB10249EA 346 EUR
ABNOMAB10249
Anti-EPCAM Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: ESA 214]
Antikörper
Mouse monoclonal antibody raised against EPCAM.

Recommended Dilutions: Immunocytochemistry: 1:50-1:100; Immunohistochemistry: 1:50-1:100; Immunofluorescence: 1:10-1:20

Type: Primary
Antigen: EPCAM
Clonality: FITC (Fluorescein Isothiocyanate)
Clone: Monoclonal
Conjugation: ESA 214
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR