Anti-MCOLN3 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB21014EA 518 EUR
ABNOPAB21014
Anti-MCOLN3 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant MCOLN3.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: MCOLN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR