Anti-APP Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB0696EA 338 EUR
ABNOPAB0696
Anti-APP Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of APP.

  • Recommended Dilutions: Immunohistochemistry: 1:500-1:3000

Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR