Anti-IARS2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB21717EA 518 EUR
ABNOPAB21717
Anti-IARS2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant IARS2.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: IARS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR