Anti-C9 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB22308EA 518 EUR
ABNOPAB22308
Anti-C9 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant C9.

Recommended Dilutions: Immunohistochemistry: 1:2500-1:5000

Type: Primary
Antigen: C9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR