Anti-HBcAg Monoclonal Antibody [clone: 13A9]

Lieferant: Abnova
ABNOMAB4774EA 643 EUR
ABNOMAB4774
Anti-HBcAg Monoclonal Antibody [clone: 13A9]
Antikörper
Mouse monoclonal antibody raised against recombinant hepatitis B core antigen.

Type: Primary
Antigen: HBcAg
Clonality: Monoclonal
Clone: 13A9
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR