Anti-SLC35D1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB13374EA 518 EUR
ABNOPAB13374
Anti-SLC35D1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of SLC35D1.

  • Recommended Dilutions: Western Blot: 1-2 μg/ml

Type: Primary
Antigen: SLC35D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR