Anti-SNPH Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB13375EA 518 EUR
ABNOPAB13375
Anti-SNPH Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of SNPH.

Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: SNPH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR