Anti-TMEM207 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB20712EA 568 EUR
ABNOPAB20712
Anti-TMEM207 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against recombinant TMEM207.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500

Type: Primary
Antigen: TMEM207
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR