Anti-BRCA1 Mouse Monoclonal Antibody [clone: KEN]

Lieferant: Abnova
ABNOMAB2363EA 643 EUR
ABNOMAB2363
Anti-BRCA1 Mouse Monoclonal Antibody [clone: KEN]
Antikörper
Mouse monoclonal antibody raised against partial recombinant BRCA1.

Recommended Dilutions: Immunocytochemistry: 1:100; Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: BRCA1
Clonality: Monoclonal
Clone: KEN
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR