Anti-BECN1 Mouse Monoclonal Antibody [clone: 1B7]

ABNOMAB2367EA 454 EUR
ABNOMAB2367
Anti-BECN1 Mouse Monoclonal Antibody [clone: 1B7]
Mouse monoclonal antibody raised against BECN1.

Recommended Dilutions: Western Blot: 2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:50-1:100

Type: Primary
Antigen: BECN1
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine, Chicken, Horse, Human, Mouse, Pig, Primates, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR