Anti-BAP1 Mouse Monoclonal Antibody [clone: 1G8]

Lieferant: Abnova
ABNOMAB2377EA 643 EUR
ABNOMAB2377
Anti-BAP1 Mouse Monoclonal Antibody [clone: 1G8]
Antikörper
Mouse monoclonal antibody raised against full length recombinant BAP1.

Recommended Dilutions: Immunocytochemistry: 1:100; Immunofluorescence: 1:50-1:200; Western Blot: 2 ?g/ml

Type: Primary
Antigen: BAP1
Clonality: Monoclonal
Clone: 1G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR