Anti-RNASEL Mouse Monoclonal Antibody [clone: 2E9]
Lieferant: Abnova
ABNOMAB2378EA 643 EUR
ABNOMAB2378
Anti-RNASEL Mouse Monoclonal Antibody [clone: 2E9]
Antikörper
Mouse monoclonal antibody raised against full length recombinant RNASEL.
Recommended Dilutions: Western Blot: 1:1000-1:4000
Type: Primary
Antigen: RNASEL
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Primates