Anti-GP6 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB13395EA 518 EUR
ABNOPAB13395
Anti-GP6 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of GP6.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: GP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR