Anti-MGMT Mouse Monoclonal Antibody [clone: MT 23.2]

Lieferant: Abnova
ABNOMAB2383EA 643 EUR
ABNOMAB2383
Anti-MGMT Mouse Monoclonal Antibody [clone: MT 23.2]
Antikörper
Mouse monoclonal antibody raised against recombinant MGMT.

Recommended Dilutions: Flow Cytometry: 1:1000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:500-1000; Western Blot: 1:1000

Type: Primary
Antigen: MGMT
Clonality: Monoclonal
Clone: MT 23.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR