Anti-NS4 Mouse Monoclonal Antibody [clone: 8A1]
Lieferant: Abnova
ABNOMAB4797EA 643 EUR
ABNOMAB4797
Anti-NS4 Mouse Monoclonal Antibody [clone: 8A1]
Antikörper
Mouse monoclonal antibody raised against partial recombinant hepatitis C virus NS-4 antigen.
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:1000
Type: Primary
Antigen: NS4
Clonality: Monoclonal
Clone: 8A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Virus