Anti-SLC25A22 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB21935EA 518 EUR
ABNOPAB21935
Anti-SLC25A22 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant SLC25A22.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: SLC25A22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR