Anti-CD44 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-85]

Lieferant: Abnova
ABNOMAB4561EA 338 EUR
ABNOMAB4561
Anti-CD44 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-85]
Antikörper
Mouse monoclonal antibody raised against native CD44.

Recommended Dilutions: Flow Cytometry: 10 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: MEM-85
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR