Anti-CLUH Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24317EA 545 EUR
ABNOPAB24317
Anti-CLUH Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant KIAA0664.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: CLUH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR