Anti-ITGA2B Mouse Monoclonal Antibody [clone: 262]

Lieferant: Abnova
ABNOMAB6395EA 448 EUR
ABNOMAB6395
Anti-ITGA2B Mouse Monoclonal Antibody [clone: 262]
Antikörper
Mouse monoclonal antibody raised against ITGA2B/ITGB3.

Recommended Dilutions: Western Blot: 10-20 ?g/ml; ELISA: 100?l of 108 washed platelets)

Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: 262
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR