Anti-SYCP1 Rabbit Polyclonal Antibody (DyLight® 649)

Lieferant: Abnova
ABNOPAB14958EA 772 EUR
ABNOPAB14958
Anti-SYCP1 Rabbit Polyclonal Antibody (DyLight® 649)
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of SYCP1.

Recommended Dilutions: Immunofluorescence: 1:300-1:750

Type:
Antigen: SYCP1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 649
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR