Anti-AXIN1 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB6140EA 440 EUR
ABNOPAB6140
Anti-AXIN1 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of AXIN1.

Recommended Dilutions: ELISA: 1:128000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 3-5 ?g/ml

Type: Primary
Antigen: AXIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR