Anti-Dengue virus E-D3 Mouse Monoclonal Antibody [clone: 2h65j]

Lieferant: Abnova
ABNOMAB8902EA 440 EUR
ABNOMAB8902
Anti-Dengue virus E-D3 Mouse Monoclonal Antibody [clone: 2h65j]
Antikörper
Mouse monoclonal antibody raised against partial recombinant Dengue virus E-D3.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Dengue virus E-D3
Clonality: Monoclonal
Clone: 2h65j
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR