Anti-TMEM205 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB18950EA 518 EUR
ABNOPAB18950
Anti-TMEM205 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of TMEM205.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.03-0.1 ?g/ml

Type: Primary
Antigen: TMEM205
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR