Anti-Mycophenolic Polyclonal Antibody

Lieferant: Abnova
ABNOPAB8320EA 1160 EUR
ABNOPAB8320
Anti-Mycophenolic Polyclonal Antibody
Antikörper
Sheep polyclonal antibody raised against Mycophenolic.

Recommended Dilutions: Enzyme Immunoassay (0.625 ?g/ml

Type: Primary
Antigen: Mycophenolic
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR