Anti-CASP9 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB9489EA 655 EUR
ABNOPAB9489
Anti-CASP9 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of CASP9.

Recommended Dilutions: Western Blot: 0.5-4 ?g/ml; Immunoprecipitation: 10-20 ?g/ml; Immunocytochemistry: 10-20 ?g/ml

Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR