Anti-IgM µ Rabbit Monoclonal Antibody

Lieferant: Abnova
ABNOPAB9186EA 655 EUR
ABNOPAB9186
Anti-IgM µ Rabbit Monoclonal Antibody
Antikörper
Rabbit anti-dog IgG recognizes dog IgM mu heavy chain. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.

Type: Secondary
Antigen: IgM µ
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Rabbit
Isotype:
Reactivity: Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR