Anti-Human IgG Monoclonal Antibody (Unconjugated)

Lieferant: Abnova
ABNOPAB9307EA 262 EUR
ABNOPAB9307
Anti-Human IgG Monoclonal Antibody (Unconjugated)
Antikörper
Mouse anti-Human IgG recognizes Human IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host:
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR