Anti-PAK6 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB7884EA 419 EUR
ABNOPAB7884
Anti-PAK6 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of PAK6.

Recommended Dilutions: ELISA: 1:2000; Western Blot: 1:500

Type: Primary
Antigen: PAK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR