Anti-Syn1 Rabbit Polyclonal Antibody

Lieferant: Cayman Chemical
CAYM10605-1EA 476 EUR
CAYM10605-1
Anti-Syn1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Syn1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR