Anti-FKBP4 Mouse Monoclonal Antibody [clone: 10F700]

Lieferant: United States Biological
USBIF4150-16B1-100EA 619 EUR
USBIF4150-16B1-100
Anti-FKBP4 Mouse Monoclonal Antibody [clone: 10F700]
Antikörper
Recommended dilution: Western Blot:1:2000; Immunochemistry:1:250; Immunoprecipitation:1:250

Type: Primary
Antigen: FKBP4
Clonality: Monoclonal
Clone: 10F700
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Hamster, Human, Mouse, Rabbit, Rat,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR