Anti-Helicobacter pylori Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIH1840-14G-20EA 368 EUR
USBIH1840-14G-20 USBIH1840-14G-100
Anti-Helicobacter pylori Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Helicobacter pylori
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR