Anti-Interleukin 1 alpha Goat Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBII7663-01D2-25EA 324 EUR
USBII7663-01D2-25
Anti-Interleukin 1 alpha Goat Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml

Type: Primary
Antigen: Interleukin 1 alpha
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR