Anti-GSC Mouse Monoclonal Antibody [clone: 4F11]

Lieferant: OriGene
ORIGCF500087EA 786 EUR
ORIGCF500087
Anti-GSC Mouse Monoclonal Antibody [clone: 4F11]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: GSC
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR