Anti-FOXA1 Mouse Monoclonal Antibody [clone: 3C1]

Lieferant: OriGene
CF500052
ORIGCF500052EA 764 EUR
ORIGCF500052
Anti-FOXA1 Mouse Monoclonal Antibody [clone: 3C1]
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: FOXA1
Clonality: Monoclonal
Clone: 3C1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR