Anti-PPP5C Mouse Monoclonal Antibody [clone: 2G2]

Lieferant: OriGene
CF500570
ORIGCF500570EA 764 EUR
ORIGCF500570
Anti-PPP5C Mouse Monoclonal Antibody [clone: 2G2]
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: PPP5C
Clonality: Monoclonal
Clone: 2G2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR