Anti-MTMR2 Mouse Monoclonal Antibody [clone: 1F10]

Lieferant: OriGene
CF502200
ORIGCF502200EA 764 EUR
ORIGCF502200
Anti-MTMR2 Mouse Monoclonal Antibody [clone: 1F10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MTMR2
Clonality: Monoclonal
Clone: 1F10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR