Anti-OCT4 Mouse Monoclonal Antibody [clone: 9B7]

Lieferant: OriGene
CF500035
ORIGCF500035EA 764 EUR
ORIGCF500035
Anti-OCT4 Mouse Monoclonal Antibody [clone: 9B7]
Antikörper
Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: OCT4
Clonality: Monoclonal
Clone: 9B7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR