Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 2E9]

Lieferant: OriGene
CF501197
ORIGCF501197EA 764 EUR
ORIGCF501197
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 2E9]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: CYP1A2
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR