Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 7D12]

Lieferant: OriGene
CF501173
ORIGCF501173EA 764 EUR
ORIGCF501173
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 7D12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CYP1A2
Clonality: Monoclonal
Clone: 7D12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR