Anti-BSG Mouse Monoclonal Antibody [clone: 3H1]

Lieferant: OriGene
CF501163
ORIGCF501163EA 764 EUR
ORIGCF501163
Anti-BSG Mouse Monoclonal Antibody [clone: 3H1]
Antikörper
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: 3H1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR