Anti-MAPRE2 Mouse Monoclonal Antibody [clone: 7C10]

Lieferant: OriGene
ORIGCF502871EA 764 EUR
ORIGCF502871
Anti-MAPRE2 Mouse Monoclonal Antibody [clone: 7C10]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MAPRE2
Clonality: Monoclonal
Clone: 7C10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR