Anti-EXD1 Mouse Monoclonal Antibody [clone: 2G5]

Lieferant: OriGene
CF502147
ORIGCF502147EA 786 EUR
ORIGCF502147
Anti-EXD1 Mouse Monoclonal Antibody [clone: 2G5]
Antikörper
Recommended Dilutions: Western Blot: 1:200-500; Flow Cytometry: 1:100

Type: Primary
Antigen: EXD1
Clonality: Monoclonal
Clone: 2G5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR