Anti-GAS7 Mouse Monoclonal Antibody [clone: 6G5]

Lieferant: OriGene
CF501825
ORIGCF501825EA 786 EUR
ORIGCF501825
Anti-GAS7 Mouse Monoclonal Antibody [clone: 6G5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GAS7
Clonality: Monoclonal
Clone: 6G5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR