Anti-STAT5A Mouse Monoclonal Antibody [clone: 3D2]

Lieferant: OriGene
ORIGCF502741EA 764 EUR
ORIGCF502741
Anti-STAT5A Mouse Monoclonal Antibody [clone: 3D2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: 3D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR