Anti-TUBA8 Mouse Monoclonal Antibody [clone: 2G6]

Lieferant: OriGene
CF501096
ORIGCF501096EA 764 EUR
ORIGCF501096
Anti-TUBA8 Mouse Monoclonal Antibody [clone: 2G6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: TUBA8
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR