Anti-DEF8 Mouse Monoclonal Antibody [clone: 4B6]

Lieferant: OriGene
CF502669
ORIGCF502669EA 764 EUR
ORIGCF502669
Anti-DEF8 Mouse Monoclonal Antibody [clone: 4B6]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DEF8
Clonality: Monoclonal
Clone: 4B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR